5-Bromo-2-thiophenecarboxaldehyde, 97%
5-Bromo-2-thiophenecarboxaldehyde, 97%
5-Bromo-2-thiophenecarboxaldehyde, 97%
5-Bromo-2-thiophenecarboxaldehyde, 97%
Thermo Scientific Chemicals
5-Bromo-2-thiophenecarboxaldehyde, 97%
CAS: 4701-17-1 | C5H3BrOS | 191.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number FSA166650050
Price (MYR)
416.00
BOT
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
416.00
BOT
Add to cart
Specifications
Chemical Name or Material5-Bromo-2-thiophenecarboxaldehyde
CAS4701-17-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0