N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
Thermo Scientific Chemicals
N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
CAS: 7570-45-8 | C15H13NO | 223.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA166030250
Price (MYR)
793.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
793.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-Ethyl-3-carbazolecarboxaldehyde
CAS7570-45-8
CAS Min %92.0
CAS Max %100.0
Melting Point85°C to 91°C