1-(2-Pyridyl)piperazine, 98%
1-(2-Pyridyl)piperazine, 98%
1-(2-Pyridyl)piperazine, 98%
1-(2-Pyridyl)piperazine, 98%
Thermo Scientific Chemicals
1-(2-Pyridyl)piperazine, 98%
CAS: 34803-66-2 | C9H13N3 | 163.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA165900250
Price (MYR)
893.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
893.00
BOT
Add to cart
Specifications
Chemical Name or Material1-(2-Pyridyl)piperazine
CAS34803-66-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0