2,4-Dimethylbenzaldehyde, 99%
2,4-Dimethylbenzaldehyde, 99%
2,4-Dimethylbenzaldehyde, 99%
2,4-Dimethylbenzaldehyde, 99%
Thermo Scientific Chemicals
2,4-Dimethylbenzaldehyde, 99%
CAS: 15764-16-6 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA165790050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 4-Dimethylbenzaldehyde
CAS15764-16-6
CAS Min %98.5
CAS Max %100.0
Melting Point-9°C