Decyl alcohol, 98+%
Decyl alcohol, 98+%
Decyl alcohol, 98+%
Decyl alcohol, 98+%
CAS: 112-30-1 | C10H22O | 158.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA165760025
Price (MYR)
552.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
552.00
EA
Add to cart
Specifications
Chemical Name or MaterialDecyl alcohol
CAS112-30-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protec