3-Diethylamino-1-propanol, 95%
3-Diethylamino-1-propanol, 95%
3-Diethylamino-1-propanol, 95%
3-Diethylamino-1-propanol, 95%
Thermo Scientific Chemicals
3-Diethylamino-1-propanol, 95%
CAS: 622-93-5 | C7H17NO | 131.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number FSA165410500
Price (MYR)
1,488.00
G
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,488.00
G
Add to cart
Specifications
Chemical Name or Material3-Diethylamino-1-propanol
Name Note97%
CAS622-93-5
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger