Methyl myristate, 99%
Methyl myristate, 99%
Methyl myristate, 99%
Methyl myristate, 99%
Thermo Scientific Chemicals
Methyl myristate, 99%
CAS: 124-10-7 | C15H30O2 | 242.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2.5 mL
Packaging:
Glass bottle
Catalog number FSA165011000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl myristate
Name Note99%
Acidity1mg KOH/g max.
CAS124-10-7
Melting Point18.4°C