N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis
N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis
N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis
N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis
Thermo Scientific Chemicals
N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis
CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic Bottle
Catalog number FSA164791000
Price (MYR)
1,042.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,042.00
BOT
Add to cart
Specifications
Absorbance(1% aq. soln.) (1 cm cell), (1% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm
Chemical Name or MaterialN, N';-Methylenebisacrylamide
Name NoteFor electrophoresis, For biochemistry
Physical FormFine Needle-like Crystals or Crystalline Powder
Acrylic Acid0.02% max.