Heptanoyl chloride, 99%
Heptanoyl chloride, 99%
Heptanoyl chloride, 99%
Heptanoyl chloride, 99%
Thermo Scientific Chemicals
Heptanoyl chloride, 99%
CAS: 2528-61-2 | C7H13ClO | 148.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA164261000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHeptanoyl chloride
Name Note99%
CAS2528-61-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
May be corrosive to metals.