2-Indanone, 97%
2-Indanone, 97%
2-Indanone, 97%
Thermo Scientific Chemicals
2-Indanone, 97%
CAS: 615-13-4 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA164110100
Price (MYR)
572.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
572.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Indanone
Name Note97%
CAS615-13-4
CAS Min %96.0
CAS Max %100.0