Adenosine, 99+%
Adenosine, 99+%
Adenosine, 99+%
Thermo Scientific Chemicals
Adenosine, 99+%
CAS: 58-61-7 | C10H13N5O4 | 267.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA164045000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAdenosine
CAS58-61-7
Absorbance0.76 to 0.80 (A250/A260),0.14 to 0.16 (A280/A260)
Melting Point233.0°C to 238.0°C