2,4-Dichlorotoluene, 99%
2,4-Dichlorotoluene, 99%
2,4-Dichlorotoluene, 99%
2,4-Dichlorotoluene, 99%
Thermo Scientific Chemicals
2,4-Dichlorotoluene, 99%
CAS: 95-73-8 | C7H6Cl2 | 161.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA163280010
Price (MYR)
1,175.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,175.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 4-Dichlorotoluene
Name Note99%
CAS95-73-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning