4-Hydroxybenzaldehyde, 99%
4-Hydroxybenzaldehyde, 99%
4-Hydroxybenzaldehyde, 99%
4-Hydroxybenzaldehyde, 99%
Thermo Scientific Chemicals
4-Hydroxybenzaldehyde, 99%
CAS: 123-08-0 | C7H6O2 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA162772500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Hydroxybenzaldehyde
CAS123-08-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec