Tripropylamine, 98%
Tripropylamine, 98%
Tripropylamine, 98%
Thermo Scientific Chemicals
Tripropylamine, 98%
CAS: 102-69-2 | C9H21N | 143.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA162690010
Price (MYR)
458.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
458.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTripropylamine
CAS102-69-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
May cause respiratory irritation.
Harmful to aquatic life wi
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several min