2,4,4-Trimethyl-2-pentene, 98%
2,4,4-Trimethyl-2-pentene, 98%
2,4,4-Trimethyl-2-pentene, 98%
2,4,4-Trimethyl-2-pentene, 98%
Thermo Scientific Chemicals
2,4,4-Trimethyl-2-pentene, 98%
CAS: 107-40-4 | C8H16 | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number FSA162630050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 4, 4-Trimethyl-2-pentene
Name Note98%
CAS107-40-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger