D(-)-Fructose, 99%
D(-)-Fructose, 99%
D(-)-Fructose, 99%
Thermo Scientific Chemicals
D(-)-Fructose, 99%
D(-)-Fructose, 99%, C6H12O6, CAS Number-57-48-7, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, arabino-hexulose, d--fructose, d---fructose, d-levulose, fructose, d, furucton, keto-d-fructose, krystar 300, sugar, fruit, 500g, Passes test, 98.5% min, 31,321, CHEBI:48095, 50ppm max, White, 200-333-3 | CAS: 57-48-7 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number FSA161355000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD(-)-Fructose
CAS57-48-7
Melting Point103.0°C to 105.0°C
ColorWhite
EINECS Number200-333-3