2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%
Thermo Scientific Chemicals
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%
CAS: 4733-39-5 | C26H20N2 | 360.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA161340050
Price (MYR)
2,110.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,110.00
EA
Add to cart
Specifications
Chemical Name or Material2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline
CAS4733-39-5
CAS Min %97.5
CAS Max %100.0
Melting Point277°C to 285°C