trans-Stilbene, 96%, Thermo Scientific Chemicals
trans-Stilbene, 96%, Thermo Scientific Chemicals
trans-Stilbene, 96%, Thermo Scientific Chemicals
trans-Stilbene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Stilbene, 96%, Thermo Scientific Chemicals
trans-Stilbene, 96%, C14H12, CAS Number-103-30-0, bibenzylidine, trans-stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, alpha,beta-diphenylethylene, stilbene, trans-1,2-diphenylethene, bibenzal, e-stilbene, bibenzylidene, 500g, 95% min. (HPLC), 05,II,537
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA161045000
Price (MYR)
3,260.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,260.00
BOT
Add to cart
Specifications
Chemical Name or Materialtrans-Stilbene
Name Note96%
CAS103-30-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Toxic to aquatic life with long lasting effects.