1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%
Thermo Scientific Chemicals
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%
CAS: 6674-22-2 | C9H16N2 | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA160615000
Price (MYR)
1,756.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,756.00
BOT
Add to cart
Specifications
Chemical Name or Material1,8-Diazabicyclo[5.4.0]undec-7-ene
Name Note≥98%
CAS6674-22-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger