1-Adamantanamine, 96%
1-Adamantanamine, 96%
1-Adamantanamine, 96%
1-Adamantanamine, 96%
Thermo Scientific Chemicals
1-Adamantanamine, 96%
CAS: 768-94-5 | C10H17N | 151.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA160470250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Adamantanamine
CAS768-94-5
CAS Min %95
Health Hazard 1Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
CAS Max %100
View more
Applications
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.
Solubility
Soluble in organic solvents. Insoluble in water.
Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.
RUO – Research Use Only