N-(3'-Aminopropyl)-2-pyrrolidinone, 95%
N-(3'-Aminopropyl)-2-pyrrolidinone, 95%
N-(3'-Aminopropyl)-2-pyrrolidinone, 95%
N-(3'-Aminopropyl)-2-pyrrolidinone, 95%
Thermo Scientific Chemicals
N-(3'-Aminopropyl)-2-pyrrolidinone, 95%
CAS: 7663-77-6 | C7H14N2O | 142.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA159600050
Price (MYR)
433.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
433.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-(3'-Aminopropyl)-2-pyrrolidinone
CAS7663-77-6
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0