2,4-Thiazolidinedione, 99%
2,4-Thiazolidinedione, 99%
2,4-Thiazolidinedione, 99%
2,4-Thiazolidinedione, 99%
Thermo Scientific Chemicals
2,4-Thiazolidinedione, 99%
CAS: 2295-31-0 | C3H3NO2S | 117.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA159490250
Price (MYR)
520.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
520.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 4-Thiazolidinedione
CAS2295-31-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0