2-Methyl-5-nitroimidazole, 99%
2-Methyl-5-nitroimidazole, 99%
2-Methyl-5-nitroimidazole, 99%
2-Methyl-5-nitroimidazole, 99%
Thermo Scientific Chemicals
2-Methyl-5-nitroimidazole, 99%
CAS: 88054-22-2 | C4H5N3O2 | 127.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number FSA159472500
Price (MYR)
634.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
634.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Methyl-5-nitroimidazole
Name Note99%
CAS88054-22-2
CAS Min %98.5
CAS Max %100.0