4,4'-Methylenebis(cyclohexylamine), 95%
4,4'-Methylenebis(cyclohexylamine), 95%
4,4'-Methylenebis(cyclohexylamine), 95%
4,4'-Methylenebis(cyclohexylamine), 95%
Thermo Scientific Chemicals
4,4'-Methylenebis(cyclohexylamine), 95%
CAS: 1761-71-3 | C13H26N2 | 210.36 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA159342500250 gGlass bottle
Catalog number FSA159342500
Price (MYR)
1,974.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,974.00
EA
Add to cart
Specifications
Chemical Name or Material4, 4'-Methylenebis(cyclohexylamine)
CAS1761-71-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lastin
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat