2-Bromo-2-methylpropane, 96%, stabilized
2-Bromo-2-methylpropane, 96%, stabilized
2-Bromo-2-methylpropane, 96%, stabilized
2-Bromo-2-methylpropane, 96%, stabilized
Thermo Scientific Chemicals
2-Bromo-2-methylpropane, 96%, stabilized
CAS: 507-19-7 | C4H9Br | 137.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA159265000
Price (MYR)
1,443.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,443.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Bromo-2-methylpropane
CAS507-19-7
Health Hazard 1GHS Signal Word: Danger
Melting Point-20.0°C
Density1.1800g/mL