DL-Alanine, 99%
DL-Alanine, 99%
DL-Alanine, 99%
Thermo Scientific Chemicals
DL-Alanine, 99%
CAS: 302-72-7 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number FSA159095000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-Alanine
Melting Point272.0°C to 275.0°C
Name Note99%
Ammonium200ppm max.
CAS302-72-7