Diisobutylamine, 99%
Diisobutylamine, 99%
Diisobutylamine, 99%
Diisobutylamine, 99%
Thermo Scientific Chemicals
Diisobutylamine, 99%
CAS: 110-96-3 | C8H20N | 130.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA159070010
Price (MYR)
455.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
455.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiisobutylamine
CAS110-96-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Flammable liquid and vapour.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi