Thiazolyl blue tetrazolium bromide, 98%
Thiazolyl blue tetrazolium bromide, 98%
Thiazolyl blue tetrazolium bromide, 98%
Thiazolyl blue tetrazolium bromide, 98%
Thermo Scientific Chemicals
Thiazolyl blue tetrazolium bromide, 98%
CAS: 298-93-1 | C18H16BrN5S | 414.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA158992500
Price (MYR)
350.00
EA
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
350.00
EA
Add to cart
Specifications
Chemical Name or MaterialThiazolyl blue tetrazolium bromide
Name Note98%
CAS298-93-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning