Isopropyl ether, 98+%, pure, stabilized with BHT
Isopropyl ether, 98+%, pure, stabilized with BHT
Isopropyl ether, 98+%, pure, stabilized with BHT
Isopropyl ether, 98+%, pure, stabilized with BHT
Thermo Scientific Chemicals
Isopropyl ether, 98+%, pure, stabilized with BHT
CAS: 108-20-3 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number FSA158960100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 L
Packaging:
Metal Drum
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropyl ether
Acidity0.005% max.
CAS108-20-3
Health Hazard 1GHS Signal Word: Danger
CAS128-37-0