Triphenylmethanol, 97%
Triphenylmethanol, 97%
Triphenylmethanol, 97%
Triphenylmethanol, 97%
Thermo Scientific Chemicals
Triphenylmethanol, 97%
CAS: 76-84-6 | C19H16O | 260.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number FSA158912500
Price (MYR)
983.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
983.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTriphenylmethanol
CAS76-84-6
Melting Point160°C to 164°C
ColorWhite to Yellow
EINECS Number200-988-5