4-Amino-1-naphthol hydrochloride, 90%, technical
4-Amino-1-naphthol hydrochloride, 90%, technical
4-Amino-1-naphthol hydrochloride, 90%, technical
4-Amino-1-naphthol hydrochloride, 90%, technical
Thermo Scientific Chemicals
4-Amino-1-naphthol hydrochloride, 90%, technical
CAS: 5959-56-8 | C10H9NO·HCl | 195.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA158510050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Amino-1-naphthol hydrochloride
Name Notetech., 90%
CAS5959-56-8
CAS Min %88.0
Health Hazard 1GHS Signal Word: Warning