Cyclohexyl isocyanide, 99%
Cyclohexyl isocyanide, 99%
Cyclohexyl isocyanide, 99%
Cyclohexyl isocyanide, 99%
Thermo Scientific Chemicals
Cyclohexyl isocyanide, 99%
CAS: 931-53-3 | C7H11N | 109.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA158400250
Price (MYR)
2,714.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,714.00
EA
Add to cart
Specifications
Chemical Name or MaterialCyclohexyl isocyanide
CAS931-53-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor