4,7-Diphenyl-1,10-phenanthroline, 99%
4,7-Diphenyl-1,10-phenanthroline, 99%
4,7-Diphenyl-1,10-phenanthroline, 99%
4,7-Diphenyl-1,10-phenanthroline, 99%
Thermo Scientific Chemicals
4,7-Diphenyl-1,10-phenanthroline, 99%
CAS: 1662-01-7 | C24H16N2 | 332.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA158320010
Price (MYR)
751.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
751.00
BOT
Add to cart
Specifications
Chemical Name or Material4, 7-Diphenyl-1, 10-phenanthroline
CAS1662-01-7
CAS Min %98.5
CAS Max %100.0
Melting Point218.0°C to 221.0°C