Trichloroethylene, 99+%, extra pure, stabilized
Trichloroethylene, 99+%, extra pure, stabilized
Trichloroethylene, 99+%, extra pure, stabilized
Trichloroethylene, 99+%, extra pure, stabilized
Thermo Scientific Chemicals
Trichloroethylene, 99+%, extra pure, stabilized
CAS: 79-01-6 | C2HCl3 | 131.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal drum
Catalog number FSA158310250
Price (MYR)
7,406.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (MYR)
7,406.00
EA
Add to cart
Specifications
Chemical Name or MaterialTrichloroethylene
Name NoteStabilized
CAS79-01-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
May cause cancer.
Suspected of causing genetic defects.