DL-2-Methyl-1-butanol, 98%
DL-2-Methyl-1-butanol, 98%
DL-2-Methyl-1-butanol, 98%
DL-2-Methyl-1-butanol, 98%
Thermo Scientific Chemicals
DL-2-Methyl-1-butanol, 98%
CAS: 137-32-6 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA158271000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-2-Methyl-1-butanol
Name Note98%
CAS137-32-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.