4,4'-Isopropylidenediphenol, 97%
4,4'-Isopropylidenediphenol, 97%
4,4'-Isopropylidenediphenol, 97%
4,4'-Isopropylidenediphenol, 97%
Thermo Scientific Chemicals
4,4'-Isopropylidenediphenol, 97%
CAS: 80-05-7 | C15H16O2 | 228.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA158245000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material4,4′-Isopropylidenediphenol
Name Note97%
CAS80-05-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger