trans-2-Hexenal, 99%
trans-2-Hexenal, 99%
trans-2-Hexenal, 99%
trans-2-Hexenal, 99%
Thermo Scientific Chemicals
trans-2-Hexenal, 99%
CAS: 6728-26-3 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA158131000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-2-Hexenal
Name Note99%
Acidity5mg KOH/g max.
CAS6728-26-3
Health Hazard 1GHS Signal Word: Danger