2-Chloropyrazine, 98%
2-Chloropyrazine, 98%
2-Chloropyrazine, 98%
2-Chloropyrazine, 98%
Thermo Scientific Chemicals
2-Chloropyrazine, 98%
CAS: 14508-49-7 | C4H3ClN2 | 114.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA158060100
Price (MYR)
371.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
371.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Chloropyrazine
CAS14508-49-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0