Tryptamine hydrochloride, 98%
Tryptamine hydrochloride, 98%
Tryptamine hydrochloride, 98%
Tryptamine hydrochloride, 98%
Thermo Scientific Chemicals
Tryptamine hydrochloride, 98%
CAS: 343-94-2 | C10H12N2·HCl | 196.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Packaging:
Glass bottle
Catalog number FSA157980100
Price (MYR)
372.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
372.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTryptamine hydrochloride
Name Note98%
CAS343-94-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: