Tetrahydrofurfurylamine, 97%
Tetrahydrofurfurylamine, 97%
Tetrahydrofurfurylamine, 97%
Tetrahydrofurfurylamine, 97%
Thermo Scientific Chemicals
Tetrahydrofurfurylamine, 97%
CAS: 4795-29-3 | C5H11NO | 101.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA157825000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrahydrofurfurylamine
Name Note97%
CAS4795-29-3
ColorColorless to Yellow
Health Hazard 1GHS Signal Word: Warning