Sarcosine, 98%
Sarcosine, 98%
Sarcosine, 98%
Thermo Scientific Chemicals
Sarcosine, 98%
CAS: 107-97-1 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA157745000
Price (MYR)
752.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
752.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSarcosine
ColorColorless or White to Yellow
CAS107-97-1
Physical FormCrystals or Powder
Quantity500 g