Pyrrole, 99%, extra pure
Pyrrole, 99%, extra pure
Pyrrole, 99%, extra pure
Pyrrole, 99%, extra pure
Thermo Scientific Chemicals
Pyrrole, 99%, extra pure
CAS: 109-97-7 | C4H5N | 67.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA157715000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPyrrole
CAS109-97-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if inhaled.
Causes serious eye damage.
Toxic if swallowed.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.