Uracil, 99+%
Uracil, 99+%
Uracil, 99+%
Thermo Scientific Chemicals
Uracil, 99+%
CAS: 66-22-8 | C4H4N2O2 | 112.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA157305000
Price (MYR)
1,957.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,957.00
EA
Add to cart
Specifications
Chemical Name or MaterialUracil
CAS66-22-8
Absorbance0.81 to 0.85 (A250/A260), 0.18 to 0.22 (A280/A260)
Melting Point335.0°C
ColorWhite to Yellow