2-Naphthol, 98%
2-Naphthol, 98%
2-Naphthol, 98%
Thermo Scientific Chemicals
2-Naphthol, 98%
CAS: 135-19-3 | C10H8O | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA156972500
Price (MYR)
293.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
293.00
EA
Add to cart
Specifications
Chemical Name or Material2-Naphthol
CAS135-19-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0