1-Methylpiperazine, 99%
1-Methylpiperazine, 99%
1-Methylpiperazine, 99%
1-Methylpiperazine, 99%
Thermo Scientific Chemicals
1-Methylpiperazine, 99%
CAS: 109-01-3 | C5H12N2 | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA156915000
Price (MYR)
1,082.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,082.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Methylpiperazine
CAS109-01-3
Health Hazard 1GHS Signal Word: Danger
Melting Point-6.0°C
Density0.9030g/mL