N,O-Bis(trimethylsilyl)acetamide, 95%
N,O-Bis(trimethylsilyl)acetamide, 95%
N,O-Bis(trimethylsilyl)acetamide, 95%
N,O-Bis(trimethylsilyl)acetamide, 95%
Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)acetamide, 95%
CAS: 10416-59-8 | C8H21NOSi2 | 203.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA156461000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN, O-Bis(trimethylsilyl)acetamide
CAS10416-59-8
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Silylation reagent
RUO – Research Use Only