7-Hydroxy-4-methylcoumarin, 97%
7-Hydroxy-4-methylcoumarin, 97%
7-Hydroxy-4-methylcoumarin, 97%
7-Hydroxy-4-methylcoumarin, 97%
Thermo Scientific Chemicals
7-Hydroxy-4-methylcoumarin, 97%
CAS: 90-33-5 | C10H8O3 | 176.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA156381000
Price (MYR)
943.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
943.00
BOT
Add to cart
Specifications
Chemical Name or Material7-Hydroxy-4-methylcoumarin
Name Note97%
CAS90-33-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning