2-Hexanol, 99%
2-Hexanol, 99%
2-Hexanol, 99%
Thermo Scientific Chemicals
2-Hexanol, 99%
CAS: 626-93-7 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA156290500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Hexanol
CAS626-93-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0