4-Fluorobenzaldehyde, 98+%
4-Fluorobenzaldehyde, 98+%
4-Fluorobenzaldehyde, 98+%
4-Fluorobenzaldehyde, 98+%
Thermo Scientific Chemicals
4-Fluorobenzaldehyde, 98+%
CAS: 459-57-4 | C7H5FO | 124.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
Packaging:
Glass bottle
Catalog number FSA156152500
Price (MYR)
1,110.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,110.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Fluorobenzaldehyde
Name Note98+%
CAS459-57-4
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning