5-Amino-1,3,4-thiadiazole-2-thiol, 98%
5-Amino-1,3,4-thiadiazole-2-thiol, 98%
5-Amino-1,3,4-thiadiazole-2-thiol, 98%
5-Amino-1,3,4-thiadiazole-2-thiol, 98%
Thermo Scientific Chemicals
5-Amino-1,3,4-thiadiazole-2-thiol, 98%
CAS: 2349-67-9 | C2H3N3S2 | 133.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA155970250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material5-Amino-1, 3, 4-thiadiazole-2-thiol
Name Note98%
CAS2349-67-9
CAS Min %97.5
CAS Max %100.0